Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Amersham™ Streptavidin-Fluor Conjugates
Shop All Cytiva Products
Click to view available options
Product Type:
Cy3-Streptavidin
Cy5-Streptavidin
Description
- High specificity reagents
Applications:
in situ hybridization, flow cytometry
Specifications
Specifications
| Formulation | This product is supplied as a lyophilized solid in Phosphate-buffered Saline (0.02M Potassium Phosphate, 0.15M NaCl) pH 7.2, containing 1%(w/v) Bovine Serum Albumin and 0.1% (w/v) Sodium Azide, reconstitute with 1mL of deionized Water, dispense into suitable aliquots and freeze, avoid repeated freeze-thaw cycles |
| For Use With (Application) | Suitable for use in the Fluorescent analysis of Biotinylated samples |
| Quantity | 1 mg |
| Product Type | Cy3-Streptavidin |
| Kit Contents | Lyophilized solid in phosphate-buffered saline (0.02M potassium phosphate, 0.15M NaCl) pH 7.2, containing 1%(w/v) bovine serum albumin and 0.1% (w/v) sodium azide, reconstitute with 1mL of deionized water, dispense into suitable aliquots and freeze, avoid repeated freeze-thaw cycles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction