Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMFR/gp78 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | AMFR/gp78 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15491620
![]() |
Novus Biologicals
NBP15491620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP154916
![]() |
Novus Biologicals
NBP154916 |
100 μL |
Each for $487.50
|
|
|||||
Description
AMFR/gp78 Polyclonal specifically detects AMFR/gp78 in Human samples. It is validated for Western Blot.Specifications
AMFR/gp78 | |
Polyclonal | |
Purified | |
RUO | |
Q6PGR1 | |
267 | |
Synthetic peptides corresponding to AMFR(autocrine motility factor receptor) The peptide sequence was selected from the C terminal of AMFR. Peptide sequence FGEVEVEPSEVEDFEARGSRFSKSADERQRMLVQRKDELLQQARKRFLNK. | |
Primary | |
73 kDa |
Western Blot | |
Unconjugated | |
Rabbit | |
Zinc Finger | |
AMF receptor, AMF receptor, isoform 1, AMF receptor, isoform 2, autocrine motility factor receptor, Autocrine motility factor receptor, isoform 2, gp78, RING finger protein 45, RNF45EC 6.3.2.- | |
AMFR | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title