Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMFR/gp78 Antibody, Novus Biologicals™
SDP

Catalog No. NBP149986 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149986 Supplier Novus Biologicals Supplier No. NBP149986
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AMFR/gp78 Polyclonal specifically detects AMFR/gp78 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen AMFR/gp78
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias AMF receptor, AMF receptor, isoform 1, AMF receptor, isoform 2, autocrine motility factor receptor, Autocrine motility factor receptor, isoform 2, gp78, RING finger protein 45, RNF45EC 6.3.2.-
Gene Symbols AMFR
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 500 and 550 of human Autocrine Motility Factor Receptor using the numbering given in entry NP_001135.3 (GeneID 267).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 267
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.