Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMFR/gp78 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB336070 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB336070 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB336070 Supplier Novus Biologicals Supplier No. NBP297986PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AMFR/gp78 Polyclonal antibody specifically detects AMFR/gp78 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen AMFR/gp78
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias AMF receptor, AMF receptor, isoform 1, AMF receptor, isoform 2, autocrine motility factor receptor, Autocrine motility factor receptor, isoform 2, gp78, RING finger protein 45, RNF45EC 6.3.2.-
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human AMFR/gp78 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 267
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.