Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AMH Monoclonal Antibody (4-1#)
Mouse Monoclonal Antibody
$243.00 - $588.50
Specifications
Antigen | AMH |
---|---|
Clone | 4-1# |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541553
|
Invitrogen™
MA541553 |
200 μL |
Each for $588.50
|
|
|||||
PIMA543968
|
Invitrogen™
MA543968 |
20 μL |
Each for $243.00
|
|
|||||
Description
AMH Monoclonal Antibody for Western Blot, ICC/IF
Anti mullerian hormone (AMH) is a member of the TGF beta superfamily. It is secreted as a homodimeric 140 kDa disulfide linked precursor that is cleaved to release the mature 30 kDa homodimer. Originally classified as a fetal testicular hormone that inhibits Mullerian duct development, AMH is expressed post natally by immature Sertoli cells, and to a lesser degree by granulosa cells. AMH plays a role in testicular differentiation and in the regulation of ovarian follicle growth.Specifications
AMH | |
1 mg/mL | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P03971 | |
268 | |
Recombinant protein Anti-Mullerian Hormone. The antigen corresponds to amino acid range 453-560 of the target protein. | |
Antibody |
4-1# | |
-20°C | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human | |
AMH | |
AMH; Anti - Mullerian hormone (Mulerian inhibiting substance); anti-Muellerian hormone; anti-Mullerian hormone; MIF; MIS; Muellerian-inhibiting factor; muellerian-inhibiting substance; Mullerian inhibiting factor; Mullerian inhibiting substance | |
AMH | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title