Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMICA/JAML Antibody (401901) [PerCP], Novus Biologicals™
SDP

Catalog No. NB372334 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372334 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372334 Supplier Novus Biologicals Supplier No. FAB34491C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AMICA/JAML Monoclonal specifically detects AMICA/JAML in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen AMICA/JAML
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 401901
Conjugate PerCP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human AMICA/JAML, Leu20-Leu275, Accession # Q86YT9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120425
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.