Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMICA/JAML Antibody (4E10), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP143309A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP143309A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP143309A32 Supplier Novus Biologicals Supplier No. NBP143309AF532
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

AMICA/JAML Monoclonal specifically detects AMICA/JAML in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen AMICA/JAML
Applications Flow Cytometry
Classification Monoclonal
Clone 40000000000
Conjugate Alexa Fluor 532
Dilution Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638
Gene Symbols JAML
Host Species Armenian Hamster
Immunogen Synthetic peptide corresponding to residues C(267) D G I L G D N F R P T Q(279) of mouse CARP.
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 120425.0
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.