Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMICA/JAML Antibody (4E10), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP143309Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143309Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143309Y Supplier Novus Biologicals Supplier No. NBP143309AF647
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

AMICA/JAML Monoclonal specifically detects AMICA/JAML in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation.

Specifications

Antigen AMICA/JAML
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 40000000000
Conjugate Alexa Fluor 647
Dilution Flow Cytometry
Gene Alias adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638
Gene Symbols AMICA1
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was JAML.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120425
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.