Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMICA/JAML Antibody (4E10), Biotin, Novus Biologicals™
SDP

Catalog No. NB358860 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358860 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358860 Supplier Novus Biologicals Supplier No. NBP143309B
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

AMICA/JAML Monoclonal antibody specifically detects AMICA/JAML in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation

Specifications

Antigen AMICA/JAML
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 4E10
Conjugate Biotin
Dilution Flow Cytometry, Immunoprecipitation
Formulation PBS
Gene Alias adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638
Host Species Armenian Hamster
Immunogen Synthetic peptide corresponding to residues C(267) D G I L G D N F R P T Q(279) of mouse CARP.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120425
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.