Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMICA/JAML Antibody (4E10) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018530 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018530 0.025 mg
NBP14330901 0.1 mg
NBP14330905 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018530 Supplier Novus Biologicals Supplier No. NBP1433090.025MG
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

AMICA/JAML Monoclonal specifically detects AMICA/JAML in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation.

Specifications

Antigen AMICA/JAML
Applications Immunoprecipitation, Flow Cytometry
Classification Monoclonal
Clone 40000000000
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500
Formulation Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Gene Alias adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638
Gene Symbols AMICA1
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was JAML.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120425
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.