Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMICA/JAML Antibody (4E10) - BSA Free, Novus Biologicals™
SDP

Armenian Hamster Monoclonal Antibody

$206.50 - $499.50

Specifications

Antigen AMICA/JAML
Clone 40000000000
Concentration 0.5 mg/mL
Dilution Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500
Applications Immunoprecipitation, Flow Cytometry
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018530
SDP
View Documents
Novus Biologicals
NBP1433090.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP14330905
SDP
View Documents
Novus Biologicals
NBP1433090.05MG
0.05 mg
Each for $315.00
Only null left
Add to Cart
 
NBP14330901
SDP
View Documents
Novus Biologicals
NBP1433090.1MG
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

AMICA/JAML Monoclonal specifically detects AMICA/JAML in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation.
Specifications

Specifications

AMICA/JAML
0.5 mg/mL
Immunoprecipitation, Flow Cytometry
Unconjugated
RUO
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
120425
The immunogen for this antibody was JAML.
Primary
Store at 4C. Do not freeze.
40000000000
Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500
Monoclonal
Armenian Hamster
Mouse
adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638
AMICA1
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.