Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMID Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB160312 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160312 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160312 Supplier Novus Biologicals Supplier No. NBP318132JF646
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AMID Polyclonal antibody specifically detects AMID in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence

Specifications

Antigen AMID
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias AMID, apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death, apoptosis-inducing factor 2, Apoptosis-inducing factor homologous mitochondrion-associated inducer of death, apoptosis-inducing factor, mitochondrion-associated, 2, Apoptosis-inducing factor-like mitochondrion-associated inducer of death, DKFZp686L1298, FLJ14497, p53-responsive gene 3 protein, PRG3apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer ofdeath, RP11-367H5.2
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 12 amino acids near the carboxyl terminus of human AMID. The immunogen is located within the last 50 amino acids of AMID.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 84883
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.