Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMIGO Antibody (S86-36), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP222413IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222413IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222413IR Supplier Novus Biologicals Supplier No. NBP222413IR

Mouse Monoclonal Antibody

AMIGO Monoclonal specifically detects AMIGO in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen AMIGO
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S86-36
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias adhesion molecule with Ig-like domain 1, ALI2, Alivin-2, AMIGO-1, AMIGOamphoterin-induced protein 1, amphoterin-induced gene and ORF, KIAA1163
Gene Symbols AMIGO1
Host Species Mouse
Immunogen Fusion protein amino acids 554-574 (cytoplasmic C-terminus) of human AMIGO-1
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 57463
Test Specificity Detects approx 60-80kDa depending on maturity/glycosylation.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.