Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aminoacetonitrile sulfate, 97%
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Aminoacetonitrile sulfate is used in organic synthesis, catalytic agent, petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Aminoacetonitrile sulfate is used in organic synthesis, catalytic agent, petrochemical additive.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from strong oxidizing agents, water/ moisture.
Aminoacetonitrile sulfate is used in organic synthesis, catalytic agent, petrochemical additive.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from strong oxidizing agents, water/ moisture.

Specifications
Specifications
Melting Point | ∼160°C (decomposition) |
Odor | Odorless |
Linear Formula | (H2NCH2CN)2·H2SO4 |
Quantity | 100 g |
UN Number | UN3439 |
Beilstein | 3916094 |
Sensitivity | Hygroscopic |
Merck Index | 14,412 |
Solubility Information | Soluble in water. |
Formula Weight | 210.21 |
Show More |
Safety and Handling
Hazard Category | H301-H315-H319-H335 |
Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S. |
EINECSNumber | 226-772-0 |
RTECSNumber | MC2100000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction