Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminoacylase/ACY1 Rabbit anti-Human, Clone: 128, Novus Biologicals™
SDP

Catalog No. NB020602 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020602 50 μg
NB020601 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB020602 Supplier Novus Biologicals Supplier No. NBP28955950UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Aminoacylase/ACY1 Monoclonal specifically detects Aminoacylase/ACY1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Aminoacylase/ACY1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 128
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500
Formulation 0.2 um filtered solution in PBS
Gene Alias ACY-1, ACY1D, acylase, aminoacylase 1, aminoacylase-1, EC 3.5.1, EC 3.5.1.14, N-acyl-L-amino-acid amidohydrolase
Host Species Rabbit
Immunogen Recombinant Human Aminoacylase/ACY1 protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 95
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.