Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aminomethanesulfonic acid, 97%

Click to view available options
Quantity:
100g
500g
13881-91-9
CH5NO3S
111.115
MFCD00008124
OBESRABRARNZJB-UHFFFAOYSA-N
methanesulfonic acid, amino,aminomethanesulphonic acid,aminomethyl sulfonic acid,aminomethylsulfonic acid,unii-ola224z482,methanesulfonic acid, 1-amino,aminomethanesulfonicacid,sodium aminomethanesulphonate,aminomethanesulfonic,aminomethanesulfoniccid
83791
aminomethanesulfonic acid
C(N)S(=O)(=O)O
13881-91-9
CH5NO3S
111.115
MFCD00008124
OBESRABRARNZJB-UHFFFAOYSA-N
methanesulfonic acid, amino,aminomethanesulphonic acid,aminomethyl sulfonic acid,aminomethylsulfonic acid,unii-ola224z482,methanesulfonic acid, 1-amino,aminomethanesulfonicacid,sodium aminomethanesulphonate,aminomethanesulfonic,aminomethanesulfoniccid
83791
aminomethanesulfonic acid
C(N)S(=O)(=O)O
Applications
Aminomethanesulfonic acid is used as a pharmaceutical intermediates.

Solubility
Soluble in water. (17 g/L) at 17°C

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.
pH 3.2
Melting Point ~180°C (decomposition)
Quantity 100g
UN Number UN2585
Beilstein 1811756
Solubility Information Soluble in water. (17g/L) at 17°C
Formula Weight 111.12
Percent Purity 97%
Chemical Name or Material Aminomethanesulfonic acid

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID

EINECSNumber : 237-649-6

TSCA : Yes

Recommended Storage : Ambient temperatures