Learn More
Aminomethyltransferase Antibody (OTI6F4), Alexa Fluor™ 700, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Aminomethyltransferase |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI6F4 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot, Immunohistochemistry |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | aminomethyltransferase, EC 2.1.2.10, GCE, GCSTaminomethyltransferase (glycine cleavage system protein T), glycine cleavage system protein T, NKHmitochondrial |
| Gene Symbols | AMT |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.