Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminopeptidase A/ENPEP Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB052892 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052892 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052892 Supplier Novus Biologicals Supplier No. NBP298221AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aminopeptidase A/ENPEP Polyclonal specifically detects Aminopeptidase A/ENPEP in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Aminopeptidase A/ENPEP
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Aminopeptidase A, APA, AP-A, CD249, CD249 antigen, Differentiation antigen gp160, EAP, EC 3.4.11, EC 3.4.11.7, glutamyl aminopeptidase, glutamyl aminopeptidase (aminopeptidase A), gp160
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Aminopeptidase A/ENPEP (P16406; Arg41-Pro945).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Neuronal Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2028
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.