Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminopeptidase LRAP/ERAP2 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB165336 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165336 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB165336 Supplier Novus Biologicals Supplier No. NBP281783AF750
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Aminopeptidase LRAP/ERAP2 Polyclonal antibody specifically detects Aminopeptidase LRAP/ERAP2 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Aminopeptidase LRAP/ERAP2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.11, EC 3.4.11.3, endoplasmic reticulum aminopeptidase 2, FLJ23633, FLJ23701, FLJ23807, Leukocyte-derived arginine aminopeptidase, LRAPL-RAPEC 3.4.11.-
Host Species Rabbit
Immunogen ERAP2 antibody was raised against a 17 amino acid peptide near the amino terminus of human ERAP2. The immunogen is located within amino acids 110 - 160 of ERAP2 . Amino Acid Squence: LQSEEDSRYMKPGKELK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 64167
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.