Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminopeptidase N/CD13 Antibody (986002), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB348141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348141 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348141 Supplier Novus Biologicals Supplier No. FAB38152MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aminopeptidase N/CD13 Monoclonal antibody specifically detects Aminopeptidase N/CD13 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF

Specifications

Antigen Aminopeptidase N/CD13
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 986002
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, AP-M, AP-N, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPNhAPN
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Aminopeptidase N/CD13, Lys69-Lys967, Accession # AAA51719
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 290
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.