Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminopeptidase N/CD13 Antibody (APN/514) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP247892B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247892B 0.2 mg
NBP247892A 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP247892B Supplier Novus Biologicals Supplier No. NBP2478920.2MG

Mouse Monoclonal Antibody

Aminopeptidase N/CD13 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen Aminopeptidase N/CD13
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Classification Monoclonal
Clone APN/514
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, AP-M, AP-N, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPNhAPN
Gene Symbols ANPEP
Host Species Mouse
Immunogen Recombinant human CD13 protein
Molecular Weight of Antigen 150 kDa
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 290
Test Specificity This MAb recognizes an extracellular epitope of an integral membrane glycoprotein of 150kDa, identified as CD13. This antigen is present on most cells of myeloid origin including granulocytes, monocytes, mast cells, and GM-progenitor cells. It is also expressed by the majority of AML, CML in myeloid blast crisis, and in a smaller fraction of lymphoid leukemias. It is absent from normal lymphocytes, platelets and erythrocytes. CD13 is also present on fibroblasts; endothelial cells, epithelial cells from renal proximal tubules and intestinal brush border, bone marrow stromal cells, osteoclasts, and cells lining bile duct canaliculi. CD13 is identical to aminopeptidase N (APN), a prominent membrane-bound metalloprotease present on the surface of intestinal brush border and renal tubules. CD13 plays a role in metabolism of biologically active peptides, in phagocytosis, and in bactericidal/tumoricidal activities. It also serves as a receptor for human coronaviruses (HCV). The lineage-restricted pattern of expression of CD13 within the hemopoietic compartment suggests that it may be important in myeloid cell differentiation.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.