Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminopeptidase N/CD13 Antibody (WM15), PerCP, Novus Biologicals™
SDP

Catalog No. NB11058720P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11058720P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11058720P Supplier Novus Biologicals Supplier No. NB11058720PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aminopeptidase N/CD13 Monoclonal specifically detects Aminopeptidase N/CD13 in Human, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen Aminopeptidase N/CD13
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay
Classification Monoclonal
Clone WM15
Conjugate PerCP
Dilution Flow Cytometry
Gene Accession No. P15144
Gene Alias alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, AP-M, AP-N, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPNhAPN
Gene Symbols ANPEP
Host Species Mouse
Immunogen Human AML cells (NM_001150.2).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 290
Test Specificity The antibody WM15 recognises the human CD13 cell surface glycoprotein, a 150 kDa molecule expressed on granulocytes, endothelial cells, epithelial cells and myeloid progenitors. HLDA III; WS Code M 213 HLDA IV; WS Code M 44 HLDA IV; WS Code M 209 HLDA V; WS Code M MA191
Target Species Human, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.