Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminopeptidase N/CD13 Rabbit anti-Mouse, Clone: 101, Novus Biologicals™
SDP

Catalog No. NB022144 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB022144 50 μg
NB022143 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB022144 Supplier Novus Biologicals Supplier No. NBP29034550UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Aminopeptidase N/CD13 Monoclonal specifically detects Aminopeptidase N/CD13 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Aminopeptidase N/CD13
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 101
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500
Formulation 0.2 um filtered solution in PBS
Gene Alias alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, AP-M, AP-N, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPNhAPN
Host Species Rabbit
Immunogen Recombinant Mouse Aminopeptidase N/CD13 protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 290
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.