Learn More
Aminopeptidase P1/XPNPEP1 Antibody (9C7), Alexa Fluor™ 405, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Aminopeptidase P1/XPNPEP1 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 9C7 |
| Conjugate | Alexa Fluor 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Aminoacylproline aminopeptidase, APP1, cytosolic, Cytosolic aminopeptidase P, EC 3.4.11.9, sAmp, xaa-Pro aminopeptidase 1, XPNPEPLX-prolyl aminopeptidase (aminopeptidase P)-like, X-prolyl aminopeptidase (aminopeptidase P) 1, soluble, X-prolyl aminopeptidase 1, soluble |
| Host Species | Mouse |
| Immunogen | Recombinant human Aminopeptidase P1/XPNPEP1 (1-623aa) purified from E. coli |
| Purification Method | Protein A purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.