Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminopeptidase P1/XPNPEP1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB104736 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB104736 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB104736 Supplier Novus Biologicals Supplier No. NBP298604C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aminopeptidase P1/XPNPEP1 Polyclonal specifically detects Aminopeptidase P1/XPNPEP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Aminopeptidase P1/XPNPEP1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias Aminoacylproline aminopeptidase, APP1, cytosolic, Cytosolic aminopeptidase P, EC 3.4.11.9, sAmp, xaa-Pro aminopeptidase 1, XPNPEPLX-prolyl aminopeptidase (aminopeptidase P)-like, X-prolyl aminopeptidase (aminopeptidase P) 1, soluble, X-prolyl aminopeptidase 1, soluble
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Aminopeptidase P1/XPNPEP1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7511
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.