Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminopeptidase PILS/ARTS1 Rabbit anti-Human, Mouse, Rat, Clone: S06-1H3, Novus Biologicals™
SDP

Catalog No. NB188188 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188188 100 μg
NB188189 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188188 Supplier Novus Biologicals Supplier No. NBP314916100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Aminopeptidase PILS/ARTS1 Monoclonal antibody specifically detects Aminopeptidase PILS/ARTS1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence

Specifications

Antigen Aminopeptidase PILS/ARTS1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone S06-1H3
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:20-1:50, Immunohistochemistry-Paraffin, Immunofluorescence
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias Adipocyte-derived leucine aminopeptidase, A-LAPALAP, Aminopeptidase PILS, ARTS1aminopeptidase regulator of TNFR1 shedding, ARTS-1PILSAP, EC 3.4.11, EC 3.4.11.-, EC 3.4.11.1, endoplasmic reticulum aminopeptidase 1, endoplasmic reticulum aminopeptidase associated with antigen processing, ERAAP1, KIAA0525APPILS, PILS-APERAAP, Puromycin-insensitive leucyl-specific aminopeptidase, Type 1 tumor necrosis factor receptor shedding aminopeptidase regulator
Host Species Rabbit
Immunogen A synthetic peptide of human Aminopeptidase PILS/ARTS1 (Uniprot#: Q9NZ08)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 51752
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.