Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonia Solution, FCC, 27-30%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
500 mL
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
Description
Spectrum™ Chemical Ammonia Solution, FCC is a solution of Ammonia in water. It has many uses, but in food production it is used primarily as a leavening agent. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| UN Number | UN2672 |
| Quantity | 20 L |
| Percent Purity | 27 to 30% |
| Grade | FCC |
| Packaging | Poly Drum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction