Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Acetate, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. m1011169012 Shop All MilliporeSigma Products
Click to view available options
Quantity:
500 g
1 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Carton
Glass Bottle
Plastic Bottle
Plastic Drum

Recommended Storage

+15°C to +25°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Melting Point 114°C
Color Colorless
pH 6.0 to 7.5
Physical Form Solid
Quantity 12 kg
Solubility Information 1480g/L
Percent Purity >96%
Packaging Plastic Drum
Density 1.17 g/3 (20 °C)
EINECSNumber 211-162-9
RTECSNumber AF3675000
Recommended Storage +15°C to +25°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.