Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium carbamate, 98%

Catalog No. p-3256875
Click to view available options
Quantity:
50 g
250 g

Ammoniating agent | CAS: 1111-78-0 | CH6N2O2 | 78.071 g/mol

Ammonium carbamate is used in the preparation of fertilizers. It acts as an ammoniating agent. It plays an important role in the formation of carbamoyl phosphate, which is essential for urea cycle and pyrimidines production. In addition to this, it is used in the preparation of trimethylsilyl carbamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1111-78-0
Molecular Formula CH6N2O2
Molecular Weight (g/mol) 78.071
MDL Number MFCD00013010
InChI Key BVCZEBOGSOYJJT-UHFFFAOYSA-N
Synonym ammonium carbamate, ammonium aminoformate, carbamic acid, monoammonium salt, carbamic acid ammonium salt, carbamic acid ammoniate, unii-i2w9615swp, carbamic acid, ammonium salt 1:1, ammoniumcarbamate, anhydride of ammonium carbonate, azanium carbamate
PubChem CID 517232
IUPAC Name azanium;carbamate
SMILES C(=O)(N)[O-].[NH4+]

Specifications

Melting Point 60° (sublimation)
Physical Form Powder
Quantity 50 g
Beilstein 3914091
Solubility Information Soluble in cold water and ammonium hydroxide. Slightly soluble in alcohol. Insoluble in acetone.
Formula Weight 78.07
Density 1.6
Chemical Name or Material Ammonium carbamate
Hazard Category H302-H318
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
EINECSNumber 214-185-2
RTECSNumber EY8575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.