Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Carbonate, ACS Reagent, Reagents

Catalog No. 01338107
Change view
Click to view available options
:
Quantity: 500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-338-107 Quantity: 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-338-107 Supplier Reagents Holdings Llc Supplier No. C1131500500A5
Only null left
Add to Cart
Add to Cart

Ammonium Carbonate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.09
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym Smelling salts
IUPAC Name carbonic acid diamine
SMILES N.N.OC(O)=O

Specifications

Boiling Point 60°C
Melting Point 58°C
Color White
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.5
Chemical Name or Material Ammonium Carbonate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.