Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Carbonate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
Wide Mouth PVC Coated Glass Bottle
Specifications
Specifications
| Quantity | 2.5 kg |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Packaging | Wide Mouth PVC Coated Glass Bottle |
| Specific Gravity | 1L = 1.50kg |
| Chemical Name or Material | Ammonium Carbonate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction