Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride-EDTA Solution, Ricca Chemical
SDP

Catalog No. 62632 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
626-32 1 L Natural Poly Bottle
626-16 500 mL Natural Poly Bottle
626-1 4 L Natural Poly Bottle
R062600020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 626-32 Supplier Ricca Chemical Company Supplier No. 62632
Add to Cart
Add to Cart

For Nitrate by the Cadmium Reduction Method

Chemical Identifiers

CAS 12125-02-9 , 1336-21-6 , 7732-18-5
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Color Colorless
Grade Laboratory
Packaging Natural Poly Bottle
pH 8.5
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Ammonium Chloride-EDTA Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.