Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Dichromate
SCI_ED

Catalog No. s25169 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Packaging:
Lab Grade - Crystal
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25169 100 g Lab Grade - Crystal
S25169A 500 g Lab Grade - Crystal
S25169B 2 kg Lab Grade - Crystal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25169

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7789-09-5
Molecular Formula Cr2H8N2O7
Molecular Weight (g/mol) 252.06
InChI Key JOSWYUNQBRPBDN-UHFFFAOYSA-P
Synonym ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481
PubChem CID 24600
IUPAC Name diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Specifications

Color Orange to Red
Quantity 100 g
Assay Percent Range 99%
Linear Formula (NH4)2Cr2O7
Formula Weight 252.06
Grade Laboratory
Packaging Lab Grade - Crystal
Density 2.15g/mL
Chemical Name or Material Ammonium Dichromate
ShelfLife 1 year
DOTInformation DOT Class 5.1, II, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.