Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium dichromate, 99.995% (metals basis)

Catalog No. p-7058753
Click to view available options
Quantity:
25 g
100 g

Catalog No. AA4318014

Supplier: Thermo Scientific Chemicals 04318014

Only null left
Add to Cart

CAS: 7789-09-5 | Cr2H8N2O7 | 252.06 g/mol

Ammonium dichromate is used in photo engraving, dye mordant, pigments, pickling, photography, process engraving, lithography, pyrotechnics, glazes for porcelain and china wares. It acts as a catalyst and a source of nitrogen in the laboratory and as the photoactive site. It is also used in the preparation of alizarin and chrome alum. Further, it is used in leather and oil industry for tanning and purification respectively. It is involved in the production of the phosphor raster television screens and other devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-09-5
Molecular Formula Cr2H8N2O7
Molecular Weight (g/mol) 252.06
MDL Number MFCD00010879
InChI Key JOSWYUNQBRPBDN-UHFFFAOYSA-P
Synonym ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481
PubChem CID 24600
IUPAC Name diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Specifications

Melting Point 170°C (decomposition)
Physical Form Crystalline
Quantity 25 g
UN Number UN1439
Merck Index 14,515
Solubility Information Soluble in water and alcohol. Insoluble in acetone.
Formula Weight 252.1
Percent Purity 99.999%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Ammonium dichromate
Show More Show Less
Hazard Category H272-H301-H312-H314-H317-H330-H334-H340-H350-H360FD-H372-H500
Hazard Statement GHS H Statement
H272-H330-H334-H340-H350-H360-H372-H314-H318-H302-H312-H317
May intensify fire
oxidizer.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: AMMONIUM DICHROMATE
EINECSNumber 232-143-1
RTECSNumber HX7650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.