Learn More
Ammonium dichromate, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 170.0°C |
| Color | Orange |
| Physical Form | Adhering Crystals |
| Quantity | 2.5 kg |
| Assay Percent Range | 98.5% min. (Iodometry) |
| Linear Formula | (NH4)2Cr2O7 |
| Merck Index | 15, 512 |
| Solubility Information | Solubility in water: 360g/L (20°C). Other solubilities: soluble in alcohol |
| Formula Weight | 252.07 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful in contact with skin. Very toxic to aquatic life with long lasting effects. May damage fertility. May damage the unborn child. May cause genetic defects. Causes damage to organs through prolonge |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. |
| EINECSNumber | 232-143-1 |
| RTECSNumber | HX7650000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.