Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium dichromate, 99%
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
| Melting Point | 170.0°C |
| Color | Orange |
| Physical Form | Adhering Crystals |
| Quantity | 100 g |
| Assay Percent Range | 98.5% min. (Iodometry) |
| Linear Formula | (NH4)2Cr2O7 |
| Merck Index | 15, 512 |
| Solubility Information | Solubility in water: 360g/L (20°C). Other solubilities: soluble in alcohol |
| Formula Weight | 252.07 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful in contact with skin. Very toxic to aquatic life with long lasting effects. May damage fertility. May damage the unborn child. May cause genetic defects. Causes damage to organs through prolonge |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. |
| EINECSNumber | 232-143-1 |
| RTECSNumber | HX7650000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction