Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Dichromate, ACS Reagent, Reagents

Catalog No. 01338103
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-103 100 g
01-338-102 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-338-103 Supplier Reagents Holdings Llc Supplier No. C1133500100C2

Due to product restrictions, please to purchase or view availability for this product.

Ammonium Dichromate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7789-09-5
Molecular Formula Cr2H8N2O7
Molecular Weight (g/mol) 252.06
InChI Key JOSWYUNQBRPBDN-UHFFFAOYSA-P
Synonym Ammonium Bichromate, Diammonium Chromate
IUPAC Name diammonium [(oxidodioxochromio)oxy]chromiumoylolate
SMILES [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

Specifications

Melting Point >180°C
Color Orange/Red
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.155
Chemical Name or Material Ammonium Dichromate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.