Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyl-p-phenylenediamine dihydrochloride, 99%

Catalog No. AC204350250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

Catalog No. AC204350250

Supplier: Thermo Scientific Chemicals 204350250

Only null left
Add to Cart

CAS: 536-46-9 | C8H14Cl2N2 | 209.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-46-9
Molecular Formula C8H14Cl2N2
Molecular Weight (g/mol) 209.11
MDL Number MFCD00012991
InChI Key IAEDWDXMFDKWFU-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-phenylenediamine dihydrochloride, 4-amino-n,n-dimethylaniline dihydrochloride, n,n-dimethyl-1,4-phenylenediamine dihydrochloride, 1,4-benzenediamine, n,n-dimethyl-, dihydrochloride, 1,4-amino-n,n-dimethylaniline,dihydrochloride, n,n-dimethyl-1,4-benzenediamine dihydrochloride, n,n-dimethyl-p-phenylenediamine 2hcl, dimethyl-p-phenylenediamine dihydrochloride, n,n-dimethyl-p-phenylenediammonium dichloride, pubchem21651
PubChem CID 2724166
IUPAC Name N1,N1-dimethylbenzene-1,4-diamine dihydrochloride
SMILES Cl.Cl.CN(C)C1=CC=C(N)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 222.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2NC6H4NH2·2HCl
Quantity 25 g
Beilstein 13, 73
Fieser 01,293
Merck Index 10, 3248
Solubility Information (5% in water) Clear colorless to purple-brown
Formula Weight 209.12
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N,N-Dimethyl-p-phenylenediamine dihydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED
EINECSNumber 208-635-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.