Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hexachloroplatinate(IV), Pt 43.4% min

Catalog No. p-7049093
Click to view available options
Quantity:
1 g
5 g

Electroplating of platinum, manufacturing of platinum sponge | CAS: 16919-58-7 | Cl6H8N2Pt | 443.86 g/mol

Electroplating of platinum, manufacturing of platinum spongeAmmonium hexachloroplatinate(IV) is used in electroplating of platinum and to make spongy platinum. It reacts with copper(II) acetate and hydrogen sulfide to prepare Cu2PtIIPt3IVS8 which finds application in diamagnetic materials and semiconductors. It is also used in platinum plating. Further, it acts as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16919-58-7
Molecular Formula Cl6H8N2Pt
Molecular Weight (g/mol) 443.86
MDL Number MFCD00010886
InChI Key PCCGQTHFYHJATL-UHFFFAOYSA-J
Synonym ammonium hexachloroplatinate iv, ammonium chloroplatinate, ammonium hexachloroplatinate, ammonium platinic chloride, ammonium platinum chloride, diammonium hexachloroplatinate 2-, diammonium platinum hexachloride, ammoniumhexachloroplatinate, epitope id:151516, platinum iv-ammonium chloride
PubChem CID 16211460
ChEBI CHEBI:59604
IUPAC Name diazanium;hexachloroplatinum(2-)
SMILES [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]

Specifications

Melting Point (decomposition)
Color Yellow
Physical Form Powder
Quantity 1 g
UN Number UN3288
Merck Index 14,548
Solubility Information Slightly soluble in water; Insoluble in alcohol,ether and concentrated hydrochloric acid.
Formula Weight 443.89
Concentration or Composition (by Analyte or Components) Pt 43.4% min.
Sensitivity Hygroscopic
Density 3.06 g/mL
Chemical Name or Material Ammonium hexachloroplatinate(IV)
Show More Show Less
Hazard Category H301-H317-H318-H334
Hazard Statement GHS H Statement
H301-H334-H372-H318-H290-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280-P285-P301+P310-P302+P352-P304+P341-P305+P351+P338-P310-P330-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 240-973-0
RTECSNumber BP5425000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.