Learn More
Ammonium hexafluorosilicate, Puratronic™, 99.999% (metals basis)
Description
Ammonium hexafluorosilicate is used as a disinfectant and analytical reagent. It finds application in etching glass, metal casting, and electroplating. It is also employed as a wood preservative agent and mothproofing agent in textiles. It plays a useful application in soldering flux. It is also used to arrest caries treatment without discoloration on demineralized primary tooth enamel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Physical Form | Crystalline |
| Quantity | 50 g |
| UN Number | UN2854 |
| Merck Index | 14,527 |
| Solubility Information | Slightly soluble in water. Insoluble in alcohol and acetone. |
| Formula Weight | 178.14 |
| Percent Purity | 99.999% |
| Grade | Puratronic™ |
| Assay | (metals basis) |
| Show More |
Safety and Handling
| Hazard Category | H301+H311+H331 |
| Hazard Statement | GHS H Statement H301-H311-H331 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: AMMONIUM FLUOROSILICATE |
| EINECSNumber | 240-968-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.