Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 10% w/v as NH3, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88489 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
500 mL

Chemical Identifiers

CAS 1336-21-6, 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
IUPAC Name amine hydrate
SMILES N.O

Specifications

Concentration 10% w/v
Quantity 500 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.