Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Hydroxide, 28.0% to 30.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
2.5 L
4 L
Packaging:
Clear Glass Bottle
Pressure Bottom Poly Bottle
Specifications
Specifications
| Quantity | 2.5 L |
| Formula Weight | 35.05 |
| Percent Purity | 28.0 to 30.0% |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Packaging | Clear Glass Bottle |
| Specific Gravity | 1L = 0.90kg |
| Chemical Name or Material | Ammonium Hydroxide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction