Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Hydroxide, 5% v/v, Reagents
Click to view available options
:
Quantity: 4 L
Specifications
Specifications
| pH | >11.5 |
| Color | Colorless |
| Boiling Point | 81°C |
| Quantity | 4 L |
| Percent Purity | 5% |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 0.995 |
| Physical Form | Liquid |
| Chemical Name or Material | Ammonium Hydroxide, 5% v/v |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction