Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 5% w/w as NH3, Reagents

Catalog No. 01336073
Change view
Click to view available options
:
Quantity: 200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-336-073 Quantity: 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-073 Supplier Reagents Holdings Llc Supplier No. CA011600200E
Only null left
Add to Cart
Add to Cart

Ammonium Hydroxide, 5% w/w as NH3, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name water
SMILES O

Specifications

pH 11.86
Color Colorless
Boiling Point 81°C
Quantity 200 L
Percent Purity 5%
Grade Reagent
Packaging Poly Drum
Specific Gravity 0.977
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, 5% w/w as NH3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.