Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Hydroxide Solution, 14.8M
SCI_ED

Catalog No. p-4416680 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Reagent Grade
Reagent Grade - Safe-Cote

Ammonium Hydroxide Solution for use in science classrooms and labs.

  • Reagent grade solution suitable for lab use
  • Comes in 500 mL and 2.5 L volumes

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
Assay Percent Range < 30% w/v
Linear Formula NH4OH
Density ∽0.9 to 1.0g/mL
Quantity 500 mL
Formula Weight 35.04
Grade Reagent
Packaging Reagent Grade - Safe-Cote
Chemical Name or Material Ammonium Hydroxide Solution
ShelfLife 5 years
DOTInformation DOT Class 8, III, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.