Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium molybdate (para) tetrahydrate, ACS, 81-83% as MoO{3}

Catalog No. p-3256885
Click to view available options
Quantity:
50 g
100 g
500 g
2 kg

Ammonium molybdate (para) tetrahydrate, H24Mo7N6O24.4H2O, CAS Number-12054-85-2, Ammonium paramolybdate, 2kg, Powder, 81-83%, 2.498, 234-722-4, 1235.86 (1163.79 Anhydrous), ACS, H302-H315-H319-H335, P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c | CAS: 12054-85-2 | H24Mo7N6O24·4H2O

Ammonium molybdate (para) tetrahydrate, is used in photography,analytical lab reagent for phosphate,lead,arsenate determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-85-2
Molecular Formula H24Mo7N6O24·4H2O
MDL Number MFCD00167059
Synonym Ammonium paramolybdate

Specifications

Color Powder
Melting Point ∽190°C (decomposition)
Assay Percent Range 81-83%
Density 2.498 g/cm3
Solubility Information Soluble in water (1:2 ratio). Insoluble in alcohol
Formula Weight 1235.86 (1163.79 Anhydrous)
Grade ACS
Odor Odorless
Quantity 50 g
Physical Form Powder
Chemical Name or Material Ammonium molybdate (para) tetrahydrate, (para) tetrahydrate
Show More Show Less
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 234-722-4
RTECSNumber QA4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.