Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium molybdate(VI) tetrahydrate, 99+%, for analysis

Catalog No. p-4509574
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

Catalog No. AC205851000

Supplier: Thermo Scientific Chemicals 205851000

Only null left
Add to Cart

CAS: 12054-85-2 | H24Mo7N6O24·4H2O | 1235.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-85-2
Molecular Formula H24Mo7N6O24·4H2O
Molecular Weight (g/mol) 1235.86

Specifications

Melting Point 190.0°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99% min
Linear Formula (NH4)6Mo7O24·4H2O
Merck Index 15,53
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 1235.86
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 2.49
Density 2.4900g/mL
Chemical Name or Material Ammonium molybdate(VI) tetrahydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.