Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate, Granular, NF, 96-102%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7783-28-0
H9N2O4P
132.06
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diammonium hydrogen phosphate
[NH4+].[NH4+].OP([O-])([O-])=O
7783-28-0
H9N2O4P
132.06
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diammonium hydrogen phosphate
[NH4+].[NH4+].OP([O-])([O-])=O

A1244, 7783-28-0, (NH4)2HPO4

Spectrum™ Ammonium Phosphate, Granular, NF is used a buffering agent excipient. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 3 ppm
Percent Purity 96 to 102%
Chloride 0.0003
Packaging Fiber Drum
pH Range 7.6-8.2
Sulfate 0.0015
Quantity 45 kg