Learn More
Ammonium phosphomolybdate hydrate, Reagent Grade
Description
Ammonium phosphomolybdate hydrate is used to prepare aluminosilicate-supported heteropolyacid catalysts, which finds application in the dehydration of glycerol in the gas phase to get acrolein. It is used as an inorganic ion exchanger involved in the removal of cesium from liquid wastes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Color | Yellow |
| Physical Form | Powder |
| Quantity | 100 g |
| Linear Formula | (NH4)3PO4·12MoO3·xH2O (x≈3) |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 1876.35 (Anhydrous) |
| Grade | Reagent |
| Density | 3.15 g/mL |
| Chemical Name or Material | Ammonium phosphomolybdate hydrate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 234-714-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.