Learn More
Ammonium sulfate, ACS, 99.0% min
Description
In biochemistry it is used for purification of proteins via ammonium sulfate precipitation method. Ammonium sulfate is used in agriculture as fertilizer. It is a slow releasing nitrogenous fertilizer and a rich source of sulphur nutrient. It is employed as a dough conditioner in bread products. It finds use as raw materials for various chemical products such as dyes and pharmacetical products. It is a component of flame-proofing agents and fire extinguisher powder.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∼280°C (decomposition) |
| Color | White |
| Physical Form | Granules |
| Quantity | 500 g |
| Linear Formula | (NH4)2SO4 |
| Merck Index | 14,555 |
| Solubility Information | Highly soluble in water; It is insoluble in acetone,ether and ethanol; It is also insoluble in liquid ammonia. |
| IUPAC Name | sulfuric acid diamine |
| Formula Weight | 132.14 |
| Percent Purity | ≥99.0% |
| Show More |
Safety and Handling
| EINECSNumber | 231-984-1 |
| RTECSNumber | BS4500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.